missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neryl acetate, 98%
CAS: 141-12-8 | C12H20O2 | 196.29 g/mol
Chemical Identifiers
| CAS | 141-12-8 |
|---|---|
| Molecular Formula | C12H20O2 |
| Molecular Weight (g/mol) | 196.29 |
| MDL Number | MFCD00063205 |
| InChI Key | HIGQPQRQIQDZMP-FLIBITNWSA-N |
| Synonym | neryl acetate, nerol acetate, neryl ethanoate, z-3,7-dimethylocta-2,6-dien-1-yl acetate, nerol acetate 6ci, unii-of82iju18h, cis-3,7-dimethyl-2,6-octadien-1-yl acetate, cis-geranyl acetate, cis-3,7-dimethyl-2,6-octadien-1-ol acetate, of82iju18h |
| PubChem CID | 1549025 |
| IUPAC Name | [(2Z)-3,7-dimethylocta-2,6-dienyl] acetate |
| SMILES | CC(=CCCC(=CCOC(=O)C)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA1928214
|
N/A | 25 g |
N/A
|
|
|||||
|
AAA1928222
|
N/A | 100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C12H20O2 | |
| MFCD00063205 | |
| neryl acetate, nerol acetate, neryl ethanoate, z-3,7-dimethylocta-2,6-dien-1-yl acetate, nerol acetate 6ci, unii-of82iju18h, cis-3,7-dimethyl-2,6-octadien-1-yl acetate, cis-geranyl acetate, cis-3,7-dimethyl-2,6-octadien-1-ol acetate, of82iju18h | |
| [(2Z)-3,7-dimethylocta-2,6-dienyl] acetate |
Specifications
| 141-12-8 | |
| 133°C to 135°C (25 mmHg) | |
| Pleasant | |
| 1.46 | |
| MFCD00063205 | |
| neryl acetate, nerol acetate, neryl ethanoate, z-3,7-dimethylocta-2,6-dien-1-yl acetate, nerol acetate 6ci, unii-of82iju18h, cis-3,7-dimethyl-2,6-octadien-1-yl acetate, cis-geranyl acetate, cis-3,7-dimethyl-2,6-octadien-1-ol acetate, of82iju18h | |
| CC(=CCCC(=CCOC(=O)C)C)C | |
| 196.29 | |
| 196.29 | |
| Neryl acetate |
| 0.91 | |
| 98°C (208°F) | |
| C12H20O2 | |
| (CH3)2C=CHCH2CH2(CH3)C=CHCH2CO2CH3 | |
| 1722814 | |
| HIGQPQRQIQDZMP-FLIBITNWSA-N | |
| [(2Z)-3,7-dimethylocta-2,6-dienyl] acetate | |
| 1549025 | |
| 98% |
Safety and Handling
EINECSNumber : 205-459-2
RTECSNumber : RG5921000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only