accessibility menu, dialog, popup

UserName

CH3OOH3CH3CH3C(Z)

CAS RN 141-12-8

IUPAC Name: (2Z)-3,7-dimethylocta-2,6-dien-1-yl acetate
Synonyms: neryl acetate geranyl acetate (isomer mix)
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: HIGQPQRQIQDZMP-FLIBITNWSA-N
SMILES: CC(C)=CCC\C(C)=C/COC(C)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)