missing translation for 'onlineSavingsMsg'
Learn More

Neryl acetate, 98%

Catalog No. AAA1928214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1928214 25 g
AAA1928222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1928214 Supplier Thermo Scientific Chemicals Supplier No. A1928214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 141-12-8 | C12H20O2 | 196.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141-12-8
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00063205
InChI Key HIGQPQRQIQDZMP-FLIBITNWSA-N
Synonym neryl acetate, nerol acetate, neryl ethanoate, z-3,7-dimethylocta-2,6-dien-1-yl acetate, nerol acetate 6ci, unii-of82iju18h, cis-3,7-dimethyl-2,6-octadien-1-yl acetate, cis-geranyl acetate, cis-3,7-dimethyl-2,6-octadien-1-ol acetate, of82iju18h
PubChem CID 1549025
IUPAC Name [(2Z)-3,7-dimethylocta-2,6-dienyl] acetate
SMILES CC(=CCCC(=CCOC(=O)C)C)C

Specifications

Density 0.91
Boiling Point 133°C to 135°C (25 mmHg)
Flash Point 98°C (208°F)
Odor Pleasant
Refractive Index 1.46
Linear Formula (CH3)2C=CHCH2CH2(CH3)C=CHCH2CO2CH3
Quantity 25 g
Beilstein 1722814
Formula Weight 196.29
Percent Purity 98%
Chemical Name or Material Neryl acetate
Show More Show Less
EINECSNumber 205-459-2
RTECSNumber RG5921000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only