missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-Carbobenzoxy-L-lysine 98.0+%, TCI America™
$106.33 - $212.64
Chemical Identifiers
| CAS | 1155-64-2 |
|---|---|
| Molecular Formula | C14H20N2O4 |
| Molecular Weight (g/mol) | 280.32 |
| MDL Number | MFCD00002638 |
| InChI Key | CKGCFBNYQJDIGS-LBPRGKRZSA-N |
| Synonym | h-lys z-oh, n6-cbz-l-lysine, n-epsilon-carbobenzyloxy-l-lysine, n epsilon-benzyloxycarbonyl-l-lysine, n6-benzyloxycarbonyl-l-lysine, nepsilon-carbobenzoxy-l-lysine, nepsilon-carbobenzyloxy-l-lysine, l-lys cbz-oh, 2s-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 6-n-cbz-l-lysine |
| PubChem CID | 1715626 |
| IUPAC Name | (2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid |
| SMILES | N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O |
Chemical Identifiers
| C14H20N2O4 | |
| MFCD00002638 | |
| h-lys z-oh, n6-cbz-l-lysine, n-epsilon-carbobenzyloxy-l-lysine, n epsilon-benzyloxycarbonyl-l-lysine, n6-benzyloxycarbonyl-l-lysine, nepsilon-carbobenzoxy-l-lysine, nepsilon-carbobenzyloxy-l-lysine, l-lys cbz-oh, 2s-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 6-n-cbz-l-lysine | |
| (2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid |
Specifications
| 1155-64-2 | |
| C14H20N2O4 | |
| h-lys z-oh, n6-cbz-l-lysine, n-epsilon-carbobenzyloxy-l-lysine, n epsilon-benzyloxycarbonyl-l-lysine, n6-benzyloxycarbonyl-l-lysine, nepsilon-carbobenzoxy-l-lysine, nepsilon-carbobenzyloxy-l-lysine, l-lys cbz-oh, 2s-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 6-n-cbz-l-lysine | |
| N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O | |
| 280.32 | |
| 280.32 | |
| Crystalline Powder |
| White | |
| MFCD00002638 | |
| CKGCFBNYQJDIGS-LBPRGKRZSA-N | |
| (2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid | |
| 1715626 | |
| ≥98.0% (T) | |
| Nepsilon-Carbobenzoxy-L-lysine |
Safety and Handling
TSCA : No