missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Carbobenzoxy-L-lysine 98.0+%, TCI America™

Catalog No. C00345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00345G 5 g
C003425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C00345G Supplier TCI America Supplier No. C00345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1155-64-2
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00002638
InChI Key CKGCFBNYQJDIGS-LBPRGKRZSA-N
Synonym h-lys z-oh, n6-cbz-l-lysine, n-epsilon-carbobenzyloxy-l-lysine, n epsilon-benzyloxycarbonyl-l-lysine, n6-benzyloxycarbonyl-l-lysine, nepsilon-carbobenzoxy-l-lysine, nepsilon-carbobenzyloxy-l-lysine, l-lys cbz-oh, 2s-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 6-n-cbz-l-lysine
PubChem CID 1715626
IUPAC Name (2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid
SMILES N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 280.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nepsilon-Carbobenzoxy-L-lysine