missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Carbobenzoxy-L-lysine 98.0+%, TCI America™

Supplier:  TCI America C003425G

Quantity 25 g

 View more versions of this product

Catalog No. C003425G


Only null left

Description

Description

Specifications

Chemical Identifiers

1155-64-2
280.32
CKGCFBNYQJDIGS-LBPRGKRZSA-N
1715626
N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
C14H20N2O4
MFCD00002638
h-lys z-oh, n6-cbz-l-lysine, n-epsilon-carbobenzyloxy-l-lysine, n epsilon-benzyloxycarbonyl-l-lysine, n6-benzyloxycarbonyl-l-lysine, nepsilon-carbobenzoxy-l-lysine, nepsilon-carbobenzyloxy-l-lysine, l-lys cbz-oh, 2s-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 6-n-cbz-l-lysine
(2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid

Specifications

Nepsilon-Carbobenzoxy-L-lysine
White
25 g
h-lys z-oh, n6-cbz-l-lysine, n-epsilon-carbobenzyloxy-l-lysine, n epsilon-benzyloxycarbonyl-l-lysine, n6-benzyloxycarbonyl-l-lysine, nepsilon-carbobenzoxy-l-lysine, nepsilon-carbobenzyloxy-l-lysine, l-lys cbz-oh, 2s-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 6-n-cbz-l-lysine
N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
280.32
280.32
Crystalline Powder
1155-64-2
C14H20N2O4
MFCD00002638
CKGCFBNYQJDIGS-LBPRGKRZSA-N
(2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid
1715626
≥98.0% (T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions