CAS RN 31202-69-4
IUPAC Name:
(2R)-2-amino-6-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Synonyms:
(2R)-2-amino-6-[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol):
246.31
Molecular Formula:
C11H22N2O4
InChi Key:
VVQIIIAZJXTLRE-SVGMAFHSNA-N
SMILES:
CC(C)(C)OC(=O)NCCCC[C@@H](N)C(O)=O
1
–
1
of
1
results
1
Promotions Available
Nepsilon-Boc-D-lysine, 98%
Nepsilon-Boc-D-lysine, 98%
CAS: 31202-69-4 Molecular Formula: C11H22N2O4 Molecular Weight (g/mol): 246.31 MDL Number: MFCD00076958 InChI Key: VVQIIIAZJXTLRE-SVGMAFHSNA-N Synonym: h-d-lys boc-oh,r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid,n-epsilon-boc-d-lysine,d-lysine, n6-1,1-dimethylethoxy carbonyl,2r-2-amino-6-tert-butoxycarbonyl amino hexanoic acid,ambotzhaa6310,n6-tert-butoxycarbonyl-d-lysine,n-epsilon-t-butyloxycarbonyl-d-lysine PubChem CID: 7018774 IUPAC Name: (2R)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid SMILES: CC(C)(C)OC(=O)NCCCC[C@@H](N)C(O)=O