missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Acetyl-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals

$443.10 - $443.10

Chemical Identifiers

CAS 159766-56-0
Molecular Formula C23H26N2O5
Molecular Weight (g/mol) 410.47
MDL Number MFCD00077409
InChI Key HQLBYVWJOXITAM-RNZRUAGMNA-N
Synonym fmoc-lys ac-oh, fmoc-n'-acetyl-l-lysine, nalpha-fmoc-nepsilon-acetyl-l-lysine, l-lysine, n6-acetyl-n2-9h-fluoren-9-ylmethoxy carbonyl, 2s-6-acetamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoc-lys ac, fmoc-lysine ac-oh, fmoc-l-lys ac-oh, ksc005q1n, fmoc-n-epsilon-acetyl-l-lysine
PubChem CID 7018846
IUPAC Name (2S)-6-acetamido-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES CC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6215406
View Documents
Thermo Scientific Chemicals
H6215406
5 g
Each for $443.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

159766-56-0
410.47
HQLBYVWJOXITAM-RNZRUAGMNA-N
7018846
CC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
C23H26N2O5
MFCD00077409
fmoc-lys ac-oh, fmoc-n'-acetyl-l-lysine, nalpha-fmoc-nepsilon-acetyl-l-lysine, l-lysine, n6-acetyl-n2-9h-fluoren-9-ylmethoxy carbonyl, 2s-6-acetamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoc-lys ac, fmoc-lysine ac-oh, fmoc-l-lys ac-oh, ksc005q1n, fmoc-n-epsilon-acetyl-l-lysine
(2S)-6-acetamido-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid

Specifications

159766-56-0
MFCD00077409
fmoc-lys ac-oh, fmoc-n'-acetyl-l-lysine, nalpha-fmoc-nepsilon-acetyl-l-lysine, l-lysine, n6-acetyl-n2-9h-fluoren-9-ylmethoxy carbonyl, 2s-6-acetamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoc-lys ac, fmoc-lysine ac-oh, fmoc-l-lys ac-oh, ksc005q1n, fmoc-n-epsilon-acetyl-l-lysine
CC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
410.47
410.46
Nepsilon-Acetyl-Nalpha-Fmoc-L-lysine
C23H26N2O5
5 g
HQLBYVWJOXITAM-RNZRUAGMNA-N
(2S)-6-acetamido-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
7018846
98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only