accessibility menu, dialog, popup

UserName

H3CONH(S)OHONHOO

CAS RN 159766-56-0

IUPAC Name: (2S)-6-acetamido-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2S)-6-acetamido-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 410.47
Molecular Formula: C23H26N2O5
InChi Key: HQLBYVWJOXITAM-RNZRUAGMNA-N
SMILES: CC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)