missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepafenac 98.0+%, TCI America™
$285.21 - $997.27
Chemical Identifiers
| CAS | 78281-72-8 |
|---|---|
| Molecular Formula | C15H14N2O2 |
| Molecular Weight (g/mol) | 254.29 |
| MDL Number | MFCD08067732 |
| InChI Key | QEFAQIPZVLVERP-UHFFFAOYSA-N |
| Synonym | 2-Amino-3-benzoylbenzeneacetamide, 2-(2-Amino-3-benzoylphenyl)acetamide |
| PubChem CID | 151075 |
| ChEBI | CHEBI:75922 |
| IUPAC Name | 2-(2-amino-3-benzoylphenyl)acetamide |
| SMILES | NC(=O)CC1=CC=CC(C(=O)C2=CC=CC=C2)=C1N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N0932200MG
|
TCI America
N0932200MG |
200 mg |
Each for $285.21
|
|
|||||
|
N09321G
|
TCI America
N09321G |
1 g |
Each for $997.27
|
|
|||||
Chemical Identifiers
| 78281-72-8 | |
| 254.29 | |
| QEFAQIPZVLVERP-UHFFFAOYSA-N | |
| 151075 | |
| 2-(2-amino-3-benzoylphenyl)acetamide |
| C15H14N2O2 | |
| MFCD08067732 | |
| 2-Amino-3-benzoylbenzeneacetamide, 2-(2-Amino-3-benzoylphenyl)acetamide | |
| CHEBI:75922 | |
| NC(=O)CC1=CC=CC(C(=O)C2=CC=CC=C2)=C1N |
Specifications
| 78281-72-8 | |
| Yellow | |
| MFCD08067732 | |
| 2-Amino-3-benzoylbenzeneacetamide, 2-(2-Amino-3-benzoylphenyl)acetamide | |
| NC(=O)CC1=CC=CC(C(=O)C2=CC=CC=C2)=C1N | |
| 254.29 | |
| CHEBI:75922 | |
| ≥98.0% (HPLC,T) | |
| Nepafenac |
| 184°C | |
| C15H14N2O2 | |
| 200 mg | |
| QEFAQIPZVLVERP-UHFFFAOYSA-N | |
| 2-(2-amino-3-benzoylphenyl)acetamide | |
| 151075 | |
| 254.29 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CY1480710
TSCA : No
Recommended Storage : Refrigerator