accessibility menu, dialog, popup

UserName

NH2OH2NO

CAS RN 78281-72-8

IUPAC Name: 2-(2-amino-3-benzoylphenyl)acetamide
Synonyms: nepafenac amfenac amide nevanac
Molecular Weight (g/mol): 254.29
Molecular Formula: C15H14N2O2
InChi Key: QEFAQIPZVLVERP-UHFFFAOYSA-N
SMILES: NC(=O)CC1=CC=CC(C(=O)C2=CC=CC=C2)=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results