missing translation for 'onlineSavingsMsg'
Learn More

Nepafenac 98.0+%, TCI America™

Catalog No. N0932200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0932200MG 200 mg
N09321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0932200MG Supplier TCI America Supplier No. N0932200MG
Only null left

Chemical Identifiers

CAS 78281-72-8
Molecular Formula C15H14N2O2
Molecular Weight (g/mol) 254.29
MDL Number MFCD08067732
InChI Key QEFAQIPZVLVERP-UHFFFAOYSA-N
Synonym 2-Amino-3-benzoylbenzeneacetamide, 2-(2-Amino-3-benzoylphenyl)acetamide
PubChem CID 151075
ChEBI CHEBI:75922
IUPAC Name 2-(2-amino-3-benzoylphenyl)acetamide
SMILES NC(=O)CC1=CC=CC(C(=O)C2=CC=CC=C2)=C1N

Specifications

Melting Point 184°C
Color Yellow
Quantity 200 mg
Formula Weight 254.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nepafenac
RTECSNumber CY1480710
TSCA No
Recommended Storage Refrigerator