missing translation for 'onlineSavingsMsg'
Learn More

Neopentylbenzene 97.0+%, TCI America™

Catalog No. P10135ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10135ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10135ML Supplier TCI America Supplier No. P10135ML

Chemical Identifiers

CAS 1007-26-7
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00038279
InChI Key CJGXJKVMUHXVHL-UHFFFAOYSA-N
Synonym neopentylbenzene, benzene, 2,2-dimethylpropyl, neopentyl benzene, benzene, neopentyl, 2,2-dimethyl-1-propyl benzene, unii-7mhd03e509, 2,2-dimethylpropyl benzene, 1-neopentylbenzene, acmc-1bs8x
PubChem CID 13877
IUPAC Name 2,2-dimethylpropylbenzene
SMILES CC(C)(C)CC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 186°C
Quantity 5 mL
UN Number 3295
Formula Weight 148.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Neopentylbenzene