CAS RN 1007-26-7
CAS RN 1007-26-7
IUPAC Name:
(2,2-dimethylpropyl)benzene
Synonyms:
neopentyl benzene
Molecular Weight (g/mol):
148.25
Molecular Formula:
C11H16
InChi Key:
CJGXJKVMUHXVHL-UHFFFAOYSA-N
SMILES:
CC(C)(C)CC1=CC=CC=C1
1
–
1
of
1
results
1
Neopentylbenzene 97.0+%, TCI America™
CAS: 1007-26-7 Molecular Formula: C11H16 Molecular Weight (g/mol): 148.249 MDL Number: MFCD00038279 InChI Key: CJGXJKVMUHXVHL-UHFFFAOYSA-N Synonym: neopentylbenzene,benzene, 2,2-dimethylpropyl,neopentyl benzene,benzene, neopentyl,2,2-dimethyl-1-propyl benzene,unii-7mhd03e509,2,2-dimethylpropyl benzene,1-neopentylbenzene,acmc-1bs8x PubChem CID: 13877 IUPAC Name: 2,2-dimethylpropylbenzene SMILES: CC(C)(C)CC1=CC=CC=C1