missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neomycin Sulfate USP, Medisca™

$61.34 - $2,146.78
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
18048040
![]() |
N/A | 1 kg |
Each for $2,146.78
|
|
|||||
18048048
![]() |
N/A | 50 g |
Each for $199.34
|
|
|||||
18048076
![]() |
N/A | 500 g |
Each for $1,288.06
|
|
|||||
18048110
![]() |
N/A | 10 g |
Each for $61.34
|
|
|||||
18048143
![]() |
N/A | 100 g |
Each for $306.69
|
|
|||||
Description
Neomycin Sulfate USP is a white to slightly yellow powder, or cryodesiccated solid. It is odorless or practically so, and is hygroscopic.
Chemical Identifiers
| 1405-10-3 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 1405-10-3 | |
| ≥ 600 μg of Neomycin/mg |
| Powder/Solid | |
| White to Light Yellow | |
| USP |