missing translation for 'onlineSavingsMsg'
Learn More

Neomycin Sulfate USP, Medisca™
GREENER_CHOICE

Catalog No. 18048040
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048040 1 kg
18048048 50 g
18048076 500 g
18048110 10 g
18048143 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18048040 Supplier Medisca Pharmaceutique Inc Supplier No. 034809

Neomycin Sulfate USP is a white to slightly yellow powder, or cryodesiccated solid. It is odorless or practically so, and is hygroscopic.

Chemical Identifiers

CAS 1405-10-3

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder/Solid
Color White to Light Yellow
Quantity 1 kg
Percent Purity ≥ 600 μg of Neomycin/mg
Grade USP