Learn More
Neomycin Sulfate USP, Medisca™
Supplier: Medisca Pharmaceutique Inc 034801
| Quantity | 10 g |
|---|
Description
Neomycin Sulfate USP is a white to slightly yellow powder, or cryodesiccated solid. It is odorless or practically so, and is hygroscopic.
Chemical Identifiers
| 1405-10-3 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 1405-10-3 | |
| 10 g | |
| USP |
| Powder/Solid | |
| White to Light Yellow | |
| ≥ 600 μg of Neomycin/mg |