missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Negative Control oligodeoxynucleotide Mouse

Catalog No. NBP231133
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231133 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP231133 Supplier R&D Systems™ Supplier No. NBP231133

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems.

Specifications

Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Line Novus
Quantity 0.1 mg
For Use With (Application) Negative control oligo for mouse TLR9 ligand
Type Negative Control Oligodeoxynucleotide