missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nefiracetam 98.0+%, TCI America™
$279.60 - $1112.84
Chemical Identifiers
| CAS | 77191-36-7 |
|---|---|
| Molecular Formula | C14H18N2O2 |
| Molecular Weight (g/mol) | 246.31 |
| MDL Number | MFCD00209882 |
| InChI Key | NGHTXZCKLWZPGK-UHFFFAOYSA-N |
| Synonym | nefiracetam, n-2,6-dimethylphenyl-2-2-oxopyrrolidin-1-yl acetamide, translon, nefiracetam inn, dmppa, motiva, nefiracetamum inn-latin, n-2,6-dimethylphenyl-2-oxo-1-pyrrolidineacetamide, unii-1jk12gx30n, ccris 6729 |
| PubChem CID | 71157 |
| IUPAC Name | N-(2,6-dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide |
| SMILES | CC1=C(C(=CC=C1)C)NC(=O)CN2CCCC2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N0996200MG
|
TCI America
N0996200MG |
200 mg |
Each for $279.60
|
|
|||||
|
N09961G
|
TCI America
N09961G |
1 g |
Each for $1,112.84
|
|
|||||
Chemical Identifiers
| 77191-36-7 | |
| 246.31 | |
| NGHTXZCKLWZPGK-UHFFFAOYSA-N | |
| 71157 | |
| CC1=C(C(=CC=C1)C)NC(=O)CN2CCCC2=O |
| C14H18N2O2 | |
| MFCD00209882 | |
| nefiracetam, n-2,6-dimethylphenyl-2-2-oxopyrrolidin-1-yl acetamide, translon, nefiracetam inn, dmppa, motiva, nefiracetamum inn-latin, n-2,6-dimethylphenyl-2-oxo-1-pyrrolidineacetamide, unii-1jk12gx30n, ccris 6729 | |
| N-(2,6-dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide |
Specifications
| 77191-36-7 | |
| White | |
| MFCD00209882 | |
| nefiracetam, n-2,6-dimethylphenyl-2-2-oxopyrrolidin-1-yl acetamide, translon, nefiracetam inn, dmppa, motiva, nefiracetamum inn-latin, n-2,6-dimethylphenyl-2-oxo-1-pyrrolidineacetamide, unii-1jk12gx30n, ccris 6729 | |
| CC1=C(C(=CC=C1)C)NC(=O)CN2CCCC2=O | |
| 246.31 | |
| 246.31 | |
| Crystalline Powder |
| 151°C | |
| C14H18N2O2 | |
| 200 mg | |
| NGHTXZCKLWZPGK-UHFFFAOYSA-N | |
| N-(2,6-dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide | |
| 71157 | |
| ≥98.0% (N) | |
| Nefiracetam |
Safety and Handling
RTECSNumber : UX9655650
TSCA : No