missing translation for 'onlineSavingsMsg'
Learn More

Nefiracetam 98.0+%, TCI America™

Catalog No. n0996200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0996200MG 200 mg
N09961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0996200MG Supplier TCI America Supplier No. N0996200MG
Only null left

Chemical Identifiers

CAS 77191-36-7
Molecular Formula C14H18N2O2
Molecular Weight (g/mol) 246.31
MDL Number MFCD00209882
InChI Key NGHTXZCKLWZPGK-UHFFFAOYSA-N
Synonym nefiracetam, n-2,6-dimethylphenyl-2-2-oxopyrrolidin-1-yl acetamide, translon, nefiracetam inn, dmppa, motiva, nefiracetamum inn-latin, n-2,6-dimethylphenyl-2-oxo-1-pyrrolidineacetamide, unii-1jk12gx30n, ccris 6729
PubChem CID 71157
IUPAC Name N-(2,6-dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide
SMILES CC1=C(C(=CC=C1)C)NC(=O)CN2CCCC2=O

Specifications

Melting Point 151°C
Color White
Quantity 200 mg
Formula Weight 246.31
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Nefiracetam
RTECSNumber UX9655650
TSCA No