accessibility menu, dialog, popup

UserName

CAS RN 77191-36-7

IUPAC Name: N-(2,6-dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide
Synonyms: nefiracetam
Molecular Weight (g/mol): 246.31
Molecular Formula: C14H18N2O2
InChi Key: NGHTXZCKLWZPGK-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1NC(=O)CN1CCCC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results