missing translation for 'onlineSavingsMsg'
Learn More

Nefazodone Hydrochloride 98.0+%, TCI America™

Catalog No. p-7134094
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1030200MG 200 mg
N10301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N1030200MG Supplier TCI America Supplier No. N1030200MG

Chemical Identifiers

CAS 82752-99-6
Molecular Formula C25H33Cl2N5O2
Molecular Weight (g/mol) 506.47
MDL Number MFCD00935760
InChI Key DYCKFEBIOUQECE-UHFFFAOYSA-N
Synonym 2-[3-[4-(3-Chlorophenyl)-1-piperazinyl]propyl]-5-ethyl-2,4-dihydro-4-(2-phenoxyethyl)-3H-1,2,4-triazol-3-one Hydrochloride
PubChem CID 54911
ChEBI CHEBI:7495
IUPAC Name hydrogen 1-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-3-ethyl-4-(2-phenoxyethyl)-4,5-dihydro-1H-1,2,4-triazol-5-one chloride
SMILES [H+].[Cl-].CCC1=NN(CCCN2CCN(CC2)C2=CC=CC(Cl)=C2)C(=O)N1CCOC1=CC=CC=C1

Specifications

Physical Form Powder/Crystals
Color White
Melting Point 188°C
Quantity 200 mg
Formula Weight 506.47
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Nefazodone Hydrochloride
RTECSNumber XZ5483009
TSCA No