accessibility menu, dialog, popup

UserName

CAS RN 82752-99-6

IUPAC Name: hydrogen 1-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-3-ethyl-4-(2-phenoxyethyl)-4,5-dihydro-1H-1,2,4-triazol-5-one chloride
Synonyms: nefazodone hcl
Molecular Weight (g/mol): 506.47
Molecular Formula: C25H33Cl2N5O2
InChi Key: DYCKFEBIOUQECE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCC1=NN(CCCN2CCN(CC2)C2=CC=CC(Cl)=C2)C(=O)N1CCOC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results