missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ NDSB-256, Calbiochem™,

$219.00 - $747.00

Chemical Identifiers

CAS 81239-45-4
Molecular Formula C12H19NO3S
Molecular Weight (g/mol) 257.35
MDL Number MFCD00225018
InChI Key MEJASPJNLSQOAG-UHFFFAOYSA-N
Synonym Dimethylbenzylammonium Propane Sulfonate
PubChem CID 4386830
IUPAC Name 3-(benzyldimethylazaniumyl)propane-1-sulfonate
SMILES C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
4800105GM
View Documents Promotion Details
MilliporeSigma™
4800105GM
5 g Glass Bottle
Each for $219.00
Only null left
 
48001025GM
View Documents Promotion Details
MilliporeSigma™
48001025GM
25 g Glass Bottle
Each for $747.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

81239-45-4
257.35
MEJASPJNLSQOAG-UHFFFAOYSA-N
4386830
C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1
C12H19NO3S
MFCD00225018
Dimethylbenzylammonium Propane Sulfonate
3-(benzyldimethylazaniumyl)propane-1-sulfonate

Specifications

White
C12H19NO3S
Dimethylbenzylammonium Propane Sulfonate
C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1
257.35
≥99% (TLC)
81239-45-4
MFCD00225018
MEJASPJNLSQOAG-UHFFFAOYSA-N
3-(benzyldimethylazaniumyl)propane-1-sulfonate
4386830
Videos
SDS
Documents

Documents