missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ NDSB-256, Calbiochem™,

Catalog No. 4800105GM
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4800105GM 5 g Glass Bottle
48001025GM 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4800105GM Supplier MilliporeSigma™ Supplier No. 4800105GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81239-45-4
Molecular Formula C12H19NO3S
Molecular Weight (g/mol) 257.35
MDL Number MFCD00225018
InChI Key MEJASPJNLSQOAG-UHFFFAOYSA-N
Synonym Dimethylbenzylammonium Propane Sulfonate
PubChem CID 4386830
IUPAC Name 3-(benzyldimethylazaniumyl)propane-1-sulfonate
SMILES C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Packaging Glass Bottle
Percent Purity ≥99% (TLC)