missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ NDSB-195, Calbiochem™,
$233.21 - $839.17
Chemical Identifiers
| CAS | 160255-06-1 |
|---|---|
| Molecular Formula | C7H17NO3S |
| Molecular Weight (g/mol) | 195.277 |
| InChI Key | NNCRHRDBFDCWPA-UHFFFAOYSA-N |
| Synonym | Dimethylethylammonium Propane Sulfonate |
| PubChem CID | 5221296 |
| IUPAC Name | 3-[ethyl(dimethyl)azaniumyl]propane-1-sulfonate |
| SMILES | CC[N+](C)(C)CCCS(=O)(=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
4800015GM
|
MilliporeSigma™
4800015GM |
5 g | Glass Bottle |
Each for $233.21
|
|
||||
|
48000125GM
|
MilliporeSigma™
48000125GM |
25 g | Glass Bottle |
Each for $839.17
|
|
||||
Chemical Identifiers
| 160255-06-1 | |
| 195.277 | |
| Dimethylethylammonium Propane Sulfonate | |
| 3-[ethyl(dimethyl)azaniumyl]propane-1-sulfonate |
| C7H17NO3S | |
| NNCRHRDBFDCWPA-UHFFFAOYSA-N | |
| 5221296 | |
| CC[N+](C)(C)CCCS(=O)(=O)[O-] |
Specifications
| White | |
| 5 g | |
| C7H17NO3S | |
| NNCRHRDBFDCWPA-UHFFFAOYSA-N | |
| 3-[ethyl(dimethyl)azaniumyl]propane-1-sulfonate | |
| 5221296 |
| 160255-06-1 | |
| Glass Bottle | |
| Dimethylethylammonium Propane Sulfonate | |
| CC[N+](C)(C)CCCS(=O)(=O)[O-] | |
| 195.277 | |
| ≥95% (TLC) |