accessibility menu, dialog, popup

UserName

CH3NCH3H3CSOOO

CAS RN 160255-06-1

IUPAC Name: 3-(ethyldimethylazaniumyl)propane-1-sulfonate
Synonyms: ethyl dimethyl ammonio propane sulfonate
Molecular Weight (g/mol): 195.28
Molecular Formula: C7H17NO3S
InChi Key: NNCRHRDBFDCWPA-UHFFFAOYSA-N
SMILES: CC[N+](C)(C)CCCS([O-])(=O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results