missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ NDSB-195, Calbiochem™,

Supplier:  MilliporeSigma™ 4800015GM

 View more versions of this product

Catalog No. 4800015GM


Only null left

Description

Description

Specifications

Specifications

White
5 g
C7H17NO3S
NNCRHRDBFDCWPA-UHFFFAOYSA-N
3-[ethyl(dimethyl)azaniumyl]propane-1-sulfonate
5221296
160255-06-1
Glass Bottle
Dimethylethylammonium Propane Sulfonate
CC[N+](C)(C)CCCS(=O)(=O)[O-]
195.277
≥95% (TLC)

Chemical Identifiers

160255-06-1
195.277
Dimethylethylammonium Propane Sulfonate
3-[ethyl(dimethyl)azaniumyl]propane-1-sulfonate
C7H17NO3S
NNCRHRDBFDCWPA-UHFFFAOYSA-N
5221296
CC[N+](C)(C)CCCS(=O)(=O)[O-]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions