missing translation for 'onlineSavingsMsg'
Learn More

Ndelta-Benzyloxycarbonyl-L-ornithine, 98%

CAS: 3304-51-6 | C13H18N2O4 | 266.297 g/mol

$69.56 - $402.49

Chemical Identifiers

CAS 3304-51-6
Molecular Formula C13H18N2O4
Molecular Weight (g/mol) 266.297
MDL Number MFCD00038200
InChI Key VULSXQYFUHKBAN-NSHDSACASA-N
Synonym h-orn z-oh, ndelta-cbz-l-ornithine, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, n-delta-benzyloxycarbonyl-l-ornithine, 2s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, h-orn cbz-oh, ambotzhaa1227, ambotzhaa1227, a-cbz-l-ornithine, a-cbz-l-ornithine
PubChem CID 7009570
IUPAC Name (2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NCCCC(C(=O)O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6243503
View Documents
Thermo Scientific Chemicals
H6243503
1 g
Each for $69.56
Only null left
 
AAH6243506
View Documents
Thermo Scientific Chemicals
H6243506
5 g
Each for $102.74
Only null left
 
AAH6243514
View Documents
Thermo Scientific Chemicals
H6243514
25 g
Each for $402.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3304-51-6
266.297
VULSXQYFUHKBAN-NSHDSACASA-N
7009570
C1=CC=C(C=C1)COC(=O)NCCCC(C(=O)O)N
C13H18N2O4
MFCD00038200
h-orn z-oh, ndelta-cbz-l-ornithine, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, n-delta-benzyloxycarbonyl-l-ornithine, 2s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, h-orn cbz-oh, ambotzhaa1227, ambotzhaa1227, a-cbz-l-ornithine, a-cbz-l-ornithine
(2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoic acid

Specifications

248°C to 252°C
C13H18N2O4
h-orn z-oh, ndelta-cbz-l-ornithine, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, n-delta-benzyloxycarbonyl-l-ornithine, 2s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, h-orn cbz-oh, ambotzhaa1227, ambotzhaa1227, a-cbz-l-ornithine, a-cbz-l-ornithine
C1=CC=C(C=C1)COC(=O)NCCCC(C(=O)O)N
1 g
7009570
98%
Ndelta-Benzyloxycarbonyl-L-ornithine
3304-51-6
MFCD00038200
VULSXQYFUHKBAN-NSHDSACASA-N
(2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoic acid
266.297
266.29
Solid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-980-8

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only