missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ndelta-Benzyloxycarbonyl-L-ornithine, 98%
CAS: 3304-51-6 | C13H18N2O4 | 266.297 g/mol
Supplier: Thermo Scientific Chemicals H6243506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 248°C to 252°C | |
| 3304-51-6 | |
| MFCD00038200 | |
| VULSXQYFUHKBAN-NSHDSACASA-N | |
| (2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoic acid | |
| 266.297 | |
| 266.29 | |
| Solid |
| Ndelta-Benzyloxycarbonyl-L-ornithine | |
| C13H18N2O4 | |
| h-orn z-oh, ndelta-cbz-l-ornithine, n'-cbz-l-ornithine, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, ornithine, n5-phenylmethoxy carbonyl, ornithine, n5-phenylmethoxy carbonyl, h-orn cbz-oh, ambotzhaa1227, n, n, a-cbz-l-ornithine | |
| C1=CC=C(C=C1)COC(=O)NCCCC(C(=O)O)N | |
| 5 g | |
| 7009570 | |
| 98% |
Chemical Identifiers
| 3304-51-6 | |
| 266.297 | |
| VULSXQYFUHKBAN-NSHDSACASA-N | |
| 7009570 | |
| C1=CC=C(C=C1)COC(=O)NCCCC(C(=O)O)N |
| C13H18N2O4 | |
| MFCD00038200 | |
| h-orn z-oh, ndelta-cbz-l-ornithine, n'-cbz-l-ornithine, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, ornithine, n5-phenylmethoxy carbonyl, ornithine, n5-phenylmethoxy carbonyl, h-orn cbz-oh, ambotzhaa1227, n, n, a-cbz-l-ornithine | |
| (2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoic acid |
Safety and Handling
EINECSNumber : 221-980-8
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only