missing translation for 'onlineSavingsMsg'
Learn More

Ndelta-Benzyloxycarbonyl-L-ornithine, 98%

CAS: 3304-51-6 | C13H18N2O4 | 266.297 g/mol

Supplier:  Thermo Scientific Chemicals H6243514

 View more versions of this product

Catalog No. AAH6243514


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

248°C to 252°C
3304-51-6
MFCD00038200
VULSXQYFUHKBAN-NSHDSACASA-N
(2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoic acid
266.297
266.29
Solid
Ndelta-Benzyloxycarbonyl-L-ornithine
C13H18N2O4
h-orn z-oh, ndelta-cbz-l-ornithine, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, ornithine, n5-phenylmethoxy carbonyl, ornithine, n5-phenylmethoxy carbonyl, h-orn cbz-oh, h-orn cbz-oh, n, a-cbz-l-ornithine, a-cbz-l-ornithine
C1=CC=C(C=C1)COC(=O)NCCCC(C(=O)O)N
25 g
7009570
98%

Chemical Identifiers

3304-51-6
266.297
VULSXQYFUHKBAN-NSHDSACASA-N
7009570
C1=CC=C(C=C1)COC(=O)NCCCC(C(=O)O)N
C13H18N2O4
MFCD00038200
h-orn z-oh, ndelta-cbz-l-ornithine, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, ornithine, n5-phenylmethoxy carbonyl, ornithine, n5-phenylmethoxy carbonyl, h-orn cbz-oh, h-orn cbz-oh, n, a-cbz-l-ornithine, a-cbz-l-ornithine
(2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-980-8

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only