missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NativePAGE™ Bis-Tris Mini Protein Gels, 4 to 16%, 1.0 mm

Catalog No. p-7237810
Encompass
Change view
Click to view available options
Wells:
10-well
15-well
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
BN1002BOX 10-well
BN1004BOX 15-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BN1002BOX Supplier Invitrogen™ Supplier No. BN1002BOX

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

One box of 10 gels

The Invitrogen NativePAGE Bis-Tris Gel System is a precast polyacrylamide mini-gel system that provides sensitive, high-resolution analysis of native proteins and protein complexes for molecular mass estimations, and assessment of purity.

The Invitrogen NativePAGE Bis-Tris Gel System is a precast polyacrylamide mini-gel system that provides sensitive, high-resolution analysis of native proteins and protein complexes for molecular mass estimations, and assessment of purity. It is based on the blue native polyacrylamide gel electrophoresis (BN PAGE) technique developed by Schägger and von Jagow, which overcomes the limitations of traditional native electrophoresis by providing a near-neutral operating pH and detergent compatibility.

Features of the NativePAGE Bis-Tris Gel System include:
• Wide molecular weight resolving range, from 15 – 10,000 kDa
• Neutral-pH separation, which better preserves the native state of protein complexes
• Resolution of all proteins in the gel regardless of their isoelectric point (pI)
• Ability to analyze membrane-protein complexes in their native conformations
• Ability to obtain better resolution than with traditional tris-glycine native electrophoresis

How NativePAGE Bis-Tris Gels work
In SDS-PAGE, SDS functions as a charge-shift molecule that denatures proteins by conferring on them a net negative charge and enables proteins to migrate towards the anode. BN PAGE uses Coomassie G-250 as the charge-shift molecule, which binds to proteins and confers a net negative charge while maintaining the proteins in their nondenatured, native state. The near-neutral pH of the NativePAGE Bis-Tris Gel System provides maximum stability of both the proteins and the gel matrix, resulting in a highly sensitive method for analysis of native membrane-protein complexes and offering superior band resolution over traditional Tris-glycine gel systems.

For transferring proteins to a membrane, we recommend using NuPAGE Transfer Buffers for traditional wet transfer and PVDF membranes. The NuPAGE transfer buffer maintains the neutral pH environment established during electrophoresis. Nitrocellulose membranes are not compatible for blotting since the membrane will bind the Coomassie G-250 dye very tightly. Alternatively, Rapid semi-dry transfer can be done using the Invitrogen Power Blotter (PB0013) or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001) using PVDF membranes.

Order Info

Shipping Conditions: Blue ice

Specifications

Gel Percentage 4 to 16%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Recommended Applications Native
Sample Loading Volume Up to 25 μL
Separation Range 15 to 10,000 kDa
Separation Type Native
Wells 10-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Wet Ice
Storage Requirements Store at 2°C to 8°C. Do not freeze.
Length (Metric) 8 cm
Width (Metric) 8 cm
Product Line NativePAGE
For Use With (Equipment) XCell SureLock Mini-Cell
Show More Show Less
What gels can I use to separate native proteins?

The NativePAGE Invitrogen Bis-Tris Gel System is a pre-cast polyacrylamide mini gel system that provides a sensitive and high-resolution method for analyzing native membrane protein complexes, native soluble proteins, molecular mass estimations, and assessing purity of native proteins. It is based on the blue native polyacrylamide gel electrophoresis technique (BN PAGE) developed by Schagger and von Jagow.

What does it mean when bands appear to be getting narrower (or "funneling") as they progress down a protein gel?

There may be too much beta-mercaptoethanol (BME), sample buffer salts, or dithiothreitol (DTT) in your samples. If the proteins are over-reduced, they can be negatively charged and actually repel each other across the lanes causing the bands to get narrower as they progress down the gel.

Are NativePAGE gels and buffers compatible with Mini Gel tank?

Yes, NativePAGE gels are compatible with our Mini Gel tank, however there is a small variation from the original protocol.

The following protocol are for using NativePage gels with the Mini Gel Tank depending on if you are also performing a western transfer or not:
If you are NOT performing a western transfer:
1. Prepare 250 mL of each buffer (Anode and Dark Blue Cathode) per gel
2. After inserting a loaded NativePAGE gel into the Mini Gel Tank and pulling the clamp forward, fill the front cathode buffer chamber to the Fill Line with Dark Blue Cathode Buffer (~200 mL per gel)
3. Add 220 mL of Anode Buffer to the back anode buffer chamber for that gel. Start the gel run

If you are performing a western transfer:
1. Prepare 250 mL of Dark Blue Cathode Buffer, 250 mL of Light Blue Cathode Buffer, and 500 mL of Anode Buffer per gel
2. After inserting a loaded NativePAGE gel into the Mini Gel Tank and pulling the clamp forward, fill the front cathode buffer chamber to the Fill Line with Dark Blue Cathode Buffer (~200 mL per gel
3. Add 220 mL of Anode Buffer to the back anode buffer chamber for that gel
4. Start the gel run; pause the run after the dark blue dye has run ~1/3 of the way through gel
a. Pour out the buffers from the Mini Gel Tank
b. Refill the back anode buffer chamber with 220 mL of Anode Buffer per gel
c. Fill the front cathode buffer chamber to the Fill Line with Light Blue Cathode Buffer (~200 mL per gel)
5. Resume the gel run

I would like to run a NativePAGE gel. Which of your protein standards should I use?

We recommend using the NativeMark Unstained Protein Standard, Cat. No. LC0725 for native gel electrophoresis with Tris-Glycine, NuPAGE Tris-Acetate or NativePAGE gels.

My NativePAGE gel stops running halfway through the electrophoresis process. Can you please help me?

During NativePAGE runs, it is common for the current to drop below 1 mA. Most power supplies register this as a “No Load” error and automatically shut off, resulting in the stopping of the gel run. This can be bypassed in some power supplies by disabling or turning off the “Load Check” feature.

Do you have any recommendations for cleaning the XCell SureLock Mini-Cell after Blue NativePAGE?

You can try the standard recommendation for cleaning which would be washing the unit with a mild detergent and rinsing with deionized water.

After western detection, my membrane has a lot of spots. What could have gone wrong?

Here are possible causes and solutions:

- Membrane blotting pads are dirty or contaminated. Soak pads with detergent and rinse thoroughly with purified water before use. Replace pads when they become worn or discolored.
- Blocking was uneven. The incubation dish must be sufficiently big to allow thorough coverage of membrane. Shake or agitate during each step.

I am getting a lot of non-specific binding after western detection. Can you offer some tips?

Here are possible causes and solutions:

- Membrane contaminated by fingerprints or keratin proteins: Wear clean gloves at all times and use forceps when handling membranes. Always handle membranes around the edges.
- Concentrated secondary antibody used: Make sure the secondary antibody is diluted as recommended. If the background remains high, but with strong band intensity, decrease the concentration of the secondary antibody.
- Concentrated Primary antibody used: Decrease the concentration of the primary antibody.
- Affinity of the primary antibody for the protein standards: Check with the protein standard manufacturer for homologies with primary antibody.
- Insufficient removal of SDS or weakly bound proteins from membrane after blotting: Follow instructions for membrane preparation before immunodetection.
- Short blocking time or long washing time: Make sure that each step is performed for the specified amount of time.

I am unable to visualize my protein bands after western detection. What is the problem?

Here are possible causes and solutions:

- The primary antibody and secondary antibody are not compatible: Use a secondary antibody that was raised against the species in which the primary antibody was raised.
- The primary antibody is too dilute: 1) Use a more concentrated antibody solution. 2) Incubate longer (e.g., overnight) at 4 degrees C. 3) Use fresh antibody and keep in mind that each time an antibody solution is used, its effective antibody concentration decreases.
- Something in your blocking buffer interferes with binding of the primary and/or secondary antibody: Try an alternate blocking buffer ± a mild surfactant like Tween-20 (0.01-0.05% v/v). There are many blocking buffer recipes available, based on non-fat dry milk, BSA, normal serum, gelatin and mixtures of these and other materials. Note that BSA (1-5%) is considered the best blocker for nitrocellulose membranes. It is easy to check the efficacy of different blocking buffers by performing dot-blots.
- The primary antibody does not recognize the protein in the species being tested: 1) Evaluate primary antibodies by dot-blotting first to how well they react with your protein. 2) Check the immunogen sequence, if provided, and determine if it is found in your protein. 3) If no immunogen sequence is available, perform a PubMed/BLAST alignment to assess the degree of homology between your target protein and the protein against which the antibody was generated. Note that many antibodies against human proteins will also recognize the non-human primate version because there is usually a high degree of amino acid identity. In contrast, many antibodies against human proteins will not recognize the corresponding proteins from rodents (and vice versa). Remember that significant homology between sequences does not guarantee that the antibody will recognize your protein. 4) Always run the recommended positive control, if available.
- Insufficient protein is bound to the membrane or the protein of interest is not abundant enough in the sample: 1) Load at least 20-30 ?g protein per lane on your gels (as a starting point), since proteins representing less than ~0.2% of the total protein are difficult to detect on western blots. 2) Use an enrichment step to increase the concentration of the target protein. For example, prepare two nuclear lysates prior to blotting nuclear proteins or perform an immunoprecipitation (IP) prior to SDS-PAGE. 3) Reduce the volume of cell extraction buffer used to lyse your cells or tissue. 4) Be sure to use freshly prepared protease inhibitors and phosphatase inhibitors, if needed, in your protein extraction buffer. 5) Run the recommended positive control, if available.
- Poor or no transfer of the proteins to the membrane 1) Check the protein transfer efficiency with a reversible protein stain like Invitrogen Reversible Membrane Protein Stain, ponceau S, amido black or use pre-stained molecular weight standards. 2) Verify that the transfer was performed with the correct electrical polarity. 3) Remember that proteins with basic pI values (e.g., histones) and high MW may not transfer well. 4) Remember that if your target protein has a low MW (≤10 kDa), it may transfer more quickly than expected. 5) If you are using PVDF membranes, make sure to pre-soak the membrane in methanol first before soaking it in transfer buffer. Note that methanol in transfer buffer increases protein binding to nitrocellulose, but omitting methanol can increase transfer efficiency of high MW proteins. 6) Low MW proteins may pass through the 0.45 µm pores in nitrocellulose membranes, so switch to NC with 0.2 or 0.1 µm pores instead.
- Excessive washing or blocking of the membrane:- 1) Avoid over-washing the membrane. Extra washing will not allow you to visualize your protein of interest if there are other problems with your blot. 2) Avoid over-blocking by using high concentrations of the blocking buffer components or long incubation times. Too much blocking can prevent your antibodies from binding to your protein. Gelatin, in particular, can mask proteins on the blot, so avoid it, if possible. Milk can also mask proteins, so instead of using 5% milk in your blocking buffer, try using it at 0.5% instead, or remove it altogether. 3) Switch to a different blocking reagent and/or block the blot for less time.
- Using the same solution of diluted primary antibody repeatedly: Use freshly-diluted antibody for each western blot because the effective concentration of a diluted antibody decreases each time it is re-used. Also, remember that dilute solutions of antibodies are less stable and may lose their activity rapidly.
- The enzyme conjugated to your secondary antibody is not working: 1) Make a fresh dilution of your secondary antibody conjugate each time you need it. Enzymes (and antibodies) may lose activity quickly in dilute solutions. 2) Omit sodium azide in buffers if you are using HRP-conjugated antibodies. 3) Avoid high heme concentrations (from blood contamination), which can interfere with HRP-based detection. 4) Avoid using phosphate in buffers with alkaline phosphatase-antibody conjugates because phosphate inhibits enzyme activity.
- Your colorimetric or other detection reagent is old and inactive: 1) Use fresh enzyme substrate for each experiment. 2) Don't use ready-to-use substrate reagents if they have changed color on their own or if they have passed their expiration date. 3) Do not dilute substrate solutions unless instructed to do so in the product manual.

I ran my protein sample on one of your gels and the bands look non-distinct and smeary after western detection. What should I do?

Here are some suggestions:

- Make sure that the correct amount of protein is loaded per lane; loading too much protein can cause smearing.
- Bands will not be as well resolved in low percentage gels; try using a higher percentage gel.
- This may be due to the antibody being too concentrated. We recommend following the manufacturer's recommended dilution or determining the optimal antibody concentration

My NativePAGE gel stops running in the middle of the electrophoresis process. Can you please help me?

During NativePAGE electrophoresis, it is common for the current to drop below 1 mA. Most power supplies register this as a “No Load” error and automatically shut off, resulting in the stopping of the gel run. This can be bypassed in some power supplies by disabling or turning off the “Load Check” feature.

What is the difference between your NativePAGE Bis-Tris gels and NuPAGE Bis-Tris gels? Can I use the NativePAGE buffers to run native applications with the NuPAGE Bis-Tris gels?

Although the gels share some major chemical components, they are not interchangeable. NuPAGE Bis-Tris gels are optimized for denaturing and reducing conditions, and their neutral pH makes it difficult to use them for native applications as most proteins will have no charge or positive charge. Therefore, native applications with these gels are not recommended. NativePAGE Bis-Tris gels on the other hand have a different formulation that has been optimized for native electrophoresis with highest resolution. The buffers for the two types of gels should not be interchanged.

Does the iBlot Dry Blotting System work with native or native-blue gels?

Yes, please take a look at the Western Blotting NativePAGE Invitrogen Bis-Tris Gels Using the iBlot 7-Minute Blotting System Application Note (http://www.thermofisher.com/content/dam/LifeTech/migration/en/filelibrary/pdf.par.18870.file.dat/native-with-iblot-app-note-v3.pdf).

Which transfer buffer do you recommend using for blotting of NativePAGE gels?

We recommend using the NuPAGE Transfer buffer for blotting of NativePAGE gels. PVDF is the recommended blotting membrane and works well in terms of transfer and detection. Nitrocellulose is not compatible with blotting of NativePAGE gels, as the nitrocellulose membrane binds the Coomassie G-250 dye very tightly and is not compatible with alcohol-containing solutions needed to destain the membrane and fix the proteins.

What staining method do you recommend for NativePAGE gels?

NativePAGE Invitrogen Bis-Tris Gels are compatible with most of the standard Coomassie R-250 or G-250 staining formulations. The Invitrogen Colloidal Blue Staining Kit (Cat. No. LC6025) is recommended for the most sensitive Coomassie staining in NativePAGE Gels. SimplyBlue Safe Stain (Cat. No. LC6060) is not recommended for use with NativePAGE gels, due to special fixing requirements. Both the SilverQuest Silver Staining Kit (Cat. No. LC6070) and SilverXpress Silver Staining Kit (Cat. No. LC6100) are suitable for staining of NativePAGE Gels. However, for best overall results and lower background, we recommend the SilverQuest Kit as the best choice.

Can I use the Invitrogen Sharp Pre-Stained Standard with the NativePAGE system?

No, most of our protein standards (including the Invitrogen Sharp prestained and unstained standards) are already denatured and reduced, and will not resolve well in a native gel. For NativePAGE electrophoresis, we offer the NativeMARK Unstained Protein Standard (Cat. No. LC0725). It is a ready-to-use protein marker that allows for molecular weight estimation of proteins using native gel electrophoresis.

Please note that even with a native standard, molecular weight estimation in native electrophoresis is very inaccurate, as gel migration can be significantly affected by differing charge and conformation of individual proteins. For more accurate estimations, use SDS-PAGE separation or mass spectrometry analysis.

Which protein standard do you recommend using with NativePAGE gels?

We recommend using the NativeMark Unstained protein standard, Cat. No. LC0725.

Can I use NativePAGE gels to perform EMSA (electrophoretic mobility shift assays)?

NativePAGE gels have been successfully used to perform EMSA showing protein-protein interactions between two purified proteins, but we have not tested NativePAGE gels for EMSA analysis of interactions between nucleic acids (either DNA or RNA) and a protein or protein complex.

What is the advantage of using NativePAGE gels for native electrophoresis as opposed to using Tris-Glycine gels under native conditions?

Samples requiring non-ionic detergents for solubility are not compatible with traditional native Tris-Glycine PAGE because as the proteins migrate into the polyacrylamide gel, they leave behind the non-ionic detergents. In the absence of non-ionic detergent, the proteins aggregate and form vertical streaks at the top of the lane. When using blue native electrophoresis (NativePAGE gels), the Coomassie G-250 dramatically reduces aggregation to allow the resolution of membrane protein complexes not seen in the Tris-Glycine gel. In addition, compared to the operative pH of the Tris-glycine system (pH 9.3-9.5), the lower operative pH of the NativePAGE gels (pH 7.5-7.7) may help to retain the native structure and/or activity of proteins sensitive to alkaline pH.

What is the size separation range for the NativePAGE gels you offer?

The NativePAGE Invitrogen Bis-Tris Gels have a wide range of separation throughout the low- and high- molecular weight ranges:

*The NativePAGE Invitrogen 3-12% Bis-Tris Gels resolve proteins in the molecular weight range of 30-10,000 kDa.
*The NativePAGE Invitrogen 4-16% Bis-Tris Gels resolve proteins in the molecular weight range of 15-10,000 kDa.

How should I store my NativePAGE gels?

We recommend storing them at 4-25 degrees C. They should not be frozen.

What are the recommended sample loading volumes and protein loading amounts for NativePAGE gels?

The recommended sample loading volumes and protein loading amounts for the NativePAGE well formats are listed on Page 3 of the NativePAGE Invitrogen Bis-Tris Gel System manual (http://tools.thermofisher.com/content/sfs/manuals/nativepage_man.pdf).

Do your NativePAGE gels contain a stacking gel?

Our NativePAGE gels do have a stacking gel. The stacking gel for these gels is a ~1 cm region at the top of the gel where the acrylamide percentage is low (4%) and constant. Below the stacking gel, the acrylamide percentage begins to increase in the gradient portion of the gel. However, the gel buffer is the same throughout the gel. So the stacking gel in NativePAGE gels is not the same as in the Laemmli system where the stacking gel has a different pH causing a decreased ion mobility for the trailing ions. Also the entire NativePAGE gel is cast in one continuous delivery due to which no demarcation line is seen between the resolving (or gradient) portion of the gel and the stacking gel.

What is the purpose of using the NativePAGE 5% G-250 Sample Additive?

The NativePAGE 5% G-250 Sample Additive is a concentrated stock solution of Coomassie G-250 designed for use with detergent (non-ionic) containing samples prepared for NativePAGE gel electrophoresis. Native proteins normally migrate on the gel according to natural charge/pI in the pH of gel buffer, but adding the Coomassie G-250 renders an associated negative charge even to high pI proteins that would normally have a positive charge. It binds to proteins non-specifically without denaturing them. The NativePAGE 5% G-250 Sample Additive is added to the cathode buffer and, optionally, to samples as well.

What is the composition of the NativePAGE 5% G-250 Sample Additive? Does it contain a detergent?

The NativePAGE 5% G-250 Sample Additive (Cat. No. BN2004) is a concentrated stock solution of Coomassie G-250 designed for use with detergent (non-ionic) containing samples prepared for NativePAGE gel electrophoresis. The exact composition of the NativePAGE 5% G-250 Sample Additive is proprietary. It does not contain a detergent.

The protein bands in some of my gel lanes are irregular or wavy? What would have caused this problem?

This could be due to:

*Debris in the well
*High salt in the sample (make sure that the salt concentration does not exceed 50-100 mM)
*Running buffer issue
*Gel casting error

I am seeing a very wavy and uneven dye front with my samples. Can you please help me troubleshoot?

This could be due to a gel polymerization issue combined with incorrect sample preparation (final sample dilution less than 1X). Please try a different lot of the same gel and make sure that the sample is correctly prepared.

I am seeing a faint, artifact doublet band at ~60 kDa in all my lanes. This band seems to be getting darker the longer I stain the gel. What could be causing this?

Possible cause:

*Excess reducing agent (beta-mercaptoethanol)
*Skin protein contaminants (keratin)

Remedy:

*The addition of iodoacetamide to the equilibration buffer just before applying the sample to the gel has been shown to eliminate these artifact bands.
*Use new electrophoretic solutions and wear gloves when handling and loading the gel. This issue is more common when highly sensitive stains are used.

I loaded different protein samples in each well but I see the same protein band in several neighboring lanes. What could have happened?

Possible cause:

*Carry-over contamination of sample from one well into neighboring wells due to loading error
*Contaminated running buffer
*Gel casting error: malformed wells

Remedy:

*Use a gel loading tip to load wells
*Reduce the sample volume
*Do not delay while loading wells
*Do not delay after the run, as proteins can diffuse horizontally; a full well left next to an empty well would eventually contaminate the empty well over time.

My protein bands appear to be skewed or distorted. What is the problem?

Possible cause:

*Poor polymerization around sample wells
*High salt concentration in sample
*Uneven gel interface
*Excessive pressure applied to the gel plates when the gel is placed into the clamp assembly
*Uneven heating of the gel
*Insoluble material in the gel or inconsistent pore size throughout the gel
*Air bubble during the run

Remedy:

*Remove excess salt/other material by dialysis, Sephadex G-25 or any other desalting column or using an Amicon concentrator.
*Either use a cooled apparatus or reduce the current at which electrophoresis is performed.

My gel seems to be lifting off the cassette. What could be causing this?

Gel lifting off the cassette can be caused by:

*Expired gels that are degrading
*Improper storage of gels
*Too much heat accumulating during the electrophoresis run due to excessive current
*Insufficient polymerization of the polyacrylamide

I am seeing a faint shadow, or "ghost" band below a normal and expected protein band? What could be the potential issue?

Ghost bands are usually attributed to a slight lifting of the gel from the cassette, which results in the trickling down of some sample beyond its normal migration point. It then accumulates and appears as a faint second band.

My protein bands in the outer lanes of the gel show a "smiling" effect. Can you please help me troubleshoot?

"Smiling" bands may be the result of the acrylamide in the gel breaking down, leaving less of a matrix for the proteins to migrate. We recommend checking to ensure that the gels have not been used past their expiration date.

I see dumbbell or barbell shaped bands after protein electrophoresis. What could be causing this?

Barbell shaped bands are a result of loading too large of a sample volume. When a large sample volume is loaded, part of the sample tends to diffuse to the sides of the wells. When the run begins and the sample moves through the stacking portion of the gel, the sample will incompletely stack causing a slight retardation of the portion of the sample that diffused to the sides of the wells. This effect may be intensified for larger proteins, whose migration is more impeded in the low concentration acrylamide of the stacking gel. To alleviate the problem, we recommend concentrating the protein and loading a smaller volume. This gives a "thinner" starting zone.

Why do I get streaking forward or "frowning" of one of my samples on my protein gel?

Here are possible causes and solutions:

1) Sample overload: Do not overload samples
2) Addition of reducing agent that is not fresh: Reduce samples right before loading and do not use samples that have been stored in reducing agent
3)Re-oxidation of the protein during the run: Add antioxidant to the running buffer if you are running NuPAGE gels
4) Presence of highly hydrophobic regions where the protein can exclude SDS: Load the sample with 2X sample buffer instead of 1X sample buffer
5) Excess salt in the sample: Precipitate and reconstitute in lower salt buffer
6) Not enough SDS in the sample: Add SDS to the upper buffer chamber (try 0.1%, 0.2%, 0.3% and 0.4% SDS)

Which protein standard do you recommend using with gels run under native conditions?

We recommend using the NativeMark Unstained Protein Standard, Cat. No. LC0725.