missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Narrow Mouth LDPE Translucent Reagent Bottles with Polypropylene Caps
Narrow-mouth LDPE reagent bottles with polypropylene caps offers chemical resistance, leakproof security, and visibility that are ideal for safely storing and handling reagents or samples in lab and industrial settings.
Specifications
| Color | Translucent |
|---|---|
| Material | LDPE |
| No. per Pack | 12 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S000098
|
Eisco Labs
CH5397PK12 |
15 mL |
N/A
|
|
|||||
|
S000099
|
Eisco Labs
CH5398PK12 |
30 mL |
N/A
|
|
|||||
|
S000100
|
Eisco Labs
CH5399PK12 |
60 mL |
N/A
|
|
|||||
|
S000101
|
Eisco Labs
CH5400PK12 |
125 mL |
N/A
|
|
|||||
|
S000102
|
Eisco Labs
CH5401PK12 |
250 mL |
N/A
|
|
|||||
|
S000103
|
Eisco Labs
CH5402PK12 |
500 mL |
N/A
|
|
|||||
Description
- LDPE reagent bottles: Includes 12 narrow mouth translucent LDPE bottles with polypropylene caps. Perfect for storage and handling of reagents and samples in labs or industries.
- Non-sterile with chemical resistance: Crafted from LDPE for resistance to a variety of chemicals, making these bottles suitable for a range of applications, including chemical storage and transport.
- Leak-proof tested: Each bottle undergoes rigorous testing to ensure a reliable seal. Leak-proof design prevents spills during storage or transport under various conditions.
- Lab-ready design: The translucent material ensures visibility for monitoring liquid levels efficiently.
- Durable for extreme conditions: Designed to perform in challenging environments with a temperature range of -100°C to +80°C. Built for reliable use in laboratories, research facilities, and industrial applications.
Specifications
| Translucent | |
| 12 Pk. | |
| Yes | |
| Narrow | |
| Not Autoclavable |
| LDPE | |
| Non-sterile | |
| No | |
| Reagent Bottle |