missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Narrow Mouth LDPE Translucent Reagent Bottles with Polypropylene Caps

Catalog No. S000098
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
125 mL
250 mL
500 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000098 15 mL
S000099 30 mL
S000100 60 mL
S000101 125 mL
S000102 250 mL
S000103 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S000098 Supplier Eisco Labs Supplier No. CH5397PK12

Narrow-mouth LDPE reagent bottles with polypropylene caps offers chemical resistance, leakproof security, and visibility that are ideal for safely storing and handling reagents or samples in lab and industrial settings.

  • LDPE reagent bottles: Includes 12 narrow mouth translucent LDPE bottles with polypropylene caps. Perfect for storage and handling of reagents and samples in labs or industries.
  • Non-sterile with chemical resistance: Crafted from LDPE for resistance to a variety of chemicals, making these bottles suitable for a range of applications, including chemical storage and transport.
  • Leak-proof tested: Each bottle undergoes rigorous testing to ensure a reliable seal. Leak-proof design prevents spills during storage or transport under various conditions.
  • Lab-ready design: The translucent material ensures visibility for monitoring liquid levels efficiently.
  • Durable for extreme conditions: Designed to perform in challenging environments with a temperature range of -100°C to +80°C. Built for reliable use in laboratories, research facilities, and industrial applications.

Specifications

Capacity (Metric) 15 mL
Capacity (English) 0.50 oz.
Color Translucent
Diameter (Metric) Inner Neck 14 mm
Material LDPE
No. per Pack 12 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 58 mm
Product Type Reagent Bottle
Diameter (Metric) 25 mm
Autoclavable Not Autoclavable
Show More Show Less