missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Narrow Mouth Amber HDPE Reagent Bottles with Polypropylene Caps
Narrow-mouth amber HDPE reagent bottles with polypropylene caps offers UV protection, leakproof performance, and chemical resistance that are ideal for safely storing light-sensitive liquids in lab or industrial settings.
Supplier: Eisco Labs CH5377PK12
Description
- Amber Reagent Bottles: Includes narrow-mouth HDPE reagent bottles with polypropylene caps, ideal for storing and transporting light-sensitive substances.
- Protect Light-Sensitive Samples: Amber color shields UV-sensitive reagents and samples, perfect for laboratory or industrial use.
- Leak-Proof and Secure: Rigorously tested for leak resistance under high-pressure conditions to ensure safe and reliable storage.
- Excellent Chemical Resistance: HDPE construction withstands a variety of chemicals, making it a versatile choice for lab applications.
- Durable and Easy To Handle: Lightweight and translucent designed for convenience in handling and transport.
Chemical Identifiers
| Amber | |
| 12 Pk. | |
| Yes | |
| Narrow | |
| Not Autoclavable |
| HDPE | |
| Non-sterile | |
| No | |
| Reagent Bottle |
Specifications
| 125 mL | |
| Amber | |
| HDPE | |
| Non-sterile | |
| No | |
| 100 mm | |
| 52 mm |
| 4.22 oz. | |
| 17.6 mm | |
| 12 Pk. | |
| Yes | |
| Narrow | |
| Reagent Bottle | |
| Not Autoclavable |