missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Narrow Mouth Amber HDPE Reagent Bottles with Polypropylene Caps

Catalog No. p-200135786
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
125 mL
500 mL
1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000086 4 mL
S000087 8 mL
S000088 15 mL
S000089 125 mL
S000090 500 mL
S000091 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S000086 Supplier Eisco Labs Supplier No. CH5372PK12
Only null left

Narrow-mouth amber HDPE reagent bottles with polypropylene caps offers UV protection, leakproof performance, and chemical resistance that are ideal for safely storing light-sensitive liquids in lab or industrial settings.

  • Amber Reagent Bottles: Includes narrow-mouth HDPE reagent bottles with polypropylene caps, ideal for storing and transporting light-sensitive substances.
  • Protect Light-Sensitive Samples: Amber color shields UV-sensitive reagents and samples, perfect for laboratory or industrial use.
  • Leak-Proof and Secure: Rigorously tested for leak resistance under high-pressure conditions to ensure safe and reliable storage.
  • Excellent Chemical Resistance: HDPE construction withstands a variety of chemicals, making it a versatile choice for lab applications.
  • Durable and Easy To Handle: Lightweight and translucent designed for convenience in handling and transport.

Specifications

Capacity (Metric) 4 mL
Capacity (English) 0.13 oz.
Color Amber
Diameter (Metric) Inner Neck 8.3 mm
For Use With (Application) Spectral Measurement of Liquids
Material HDPE
No. per Pack 12 Pk.
Sterility Non-sterile
Width (Metric) 15.24 mm
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 41 mm
Product Type Reagent Bottle
Diameter (Metric) 16 mm
Autoclavable Not Autoclavable
Show More Show Less