missing translation for 'onlineSavingsMsg'
Learn More

Naphthol AS-BI phosphate, 93%

CAS: 1919-91-1 | C18H15BrNO6P | 452.2 g/mol

Supplier:  Thermo Scientific Chemicals 415310010

 View more versions of this product

Catalog No. AC415310010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Naphthol AS-BI phosphate
91.0
Yellow
91% min. (HPLC)
C18H15BrNO6P
1 g
Solubility in water: partly soluble
COC1=CC=CC=C1NC(=O)C2=C(C=C3C=CC(=CC3=C2)Br)OP(=O)(O)O
452.2
452.2
Crystalline Powder
1919-91-1
100.0
Authentic
Glass bottle
MFCD00004058
naphthol as-bi phosphate, naphthol as bi phosphate, 7-bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate, 2-naphthalenecarboxamide, 7-bromo-n-2-methoxyphenyl-3-phosphonooxy, naphthol as-bi phosphoric acid, naphthol as-bi phosphoric acid solution, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl oxyphosphonic acid, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl dihydrogen phosphate, 2-naphthalenecarboxamide,7-bromo-n-2-methoxyphenyl-3-phosphonooxy-, sodium salt 1:2, naphthol as-bl phosphate
HUXIAXQSTATULQ-UHFFFAOYSA-N
[6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl] dihydrogen phosphate
74715
93%

Chemical Identifiers

1919-91-1
452.2
HUXIAXQSTATULQ-UHFFFAOYSA-N
74715
COC1=CC=CC=C1NC(=O)C2=C(C=C3C=CC(=CC3=C2)Br)OP(=O)(O)O
C18H15BrNO6P
MFCD00004058
naphthol as-bi phosphate, naphthol as bi phosphate, 7-bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate, 2-naphthalenecarboxamide, 7-bromo-n-2-methoxyphenyl-3-phosphonooxy, naphthol as-bi phosphoric acid, naphthol as-bi phosphoric acid solution, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl oxyphosphonic acid, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl dihydrogen phosphate, 2-naphthalenecarboxamide,7-bromo-n-2-methoxyphenyl-3-phosphonooxy-, sodium salt 1:2, naphthol as-bl phosphate
[6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl] dihydrogen phosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 217-645-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only