accessibility menu, dialog, popup

UserName

CAS RN 1919-91-1

IUPAC Name: disodium 6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl phosphate
Synonyms: disodium 6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl phosphate
Molecular Weight (g/mol): 496.16
Molecular Formula: C18H13BrNNa2O6P
InChi Key: HESHHBAVKCAKQF-UHFFFAOYSA-L
SMILES: [Na+].[Na+].COC1=CC=CC=C1NC(=O)C1=C(OP([O-])([O-])=O)C=C2C=CC(Br)=CC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results