missing translation for 'onlineSavingsMsg'
Learn More

Naphthol AS-BI phosphate, 93%

Catalog No. p-3758541
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415311000 100 mg Glass bottle
AC415310010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC415311000 Supplier Thermo Scientific Chemicals Supplier No. 415311000
Only null left

CAS: 1919-91-1 | C18H15BrNO6P | 452.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1919-91-1
Molecular Formula C18H15BrNO6P
Molecular Weight (g/mol) 452.2
MDL Number MFCD00004058
InChI Key HUXIAXQSTATULQ-UHFFFAOYSA-N
Synonym naphthol as-bi phosphate, naphthol as bi phosphate, 7-bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate, 2-naphthalenecarboxamide, 7-bromo-n-2-methoxyphenyl-3-phosphonooxy, naphthol as-bi phosphoric acid, naphthol as-bi phosphoric acid solution, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl oxyphosphonic acid, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl dihydrogen phosphate, 2-naphthalenecarboxamide,7-bromo-n-2-methoxyphenyl-3-phosphonooxy-, sodium salt 1:2, naphthol as-bl phosphate
PubChem CID 74715
IUPAC Name [6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl] dihydrogen phosphate
SMILES COC1=CC=CC=C1NC(=O)C2=C(C=C3C=CC(=CC3=C2)Br)OP(=O)(O)O

Specifications

CAS Min % 91.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 91% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: partly soluble
Formula Weight 452.2
Percent Purity 93%
Physical Form Crystalline Powder
Chemical Name or Material Naphthol AS-BI phosphate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
EINECSNumber 217-645-

RUO – Research Use Only