Learn More
Naphthalene-2-sulfonic acid sodium salt, 98%, may cont. up to 10% residual inorganic salts and water
CAS: 532-02-5 | C10H7NaO3S | 230.21 g/mol
Supplier: Thermo Scientific Chemicals A130660B
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Naphthalene-2-sulfonic acid sodium salt | |
| may cont. up to 10% residual inorganic salts and water | |
| MFCD00064186 | |
| 3659024 | |
| sodium 2-naphthalenesulfonate, sodium naphthalene-2-sulfonate, 2-naphthalenesulfonic acid sodium salt, 2-naphthalenesulfonic acid, sodium salt, unii-r5f0ctd2oj, sodium-2-naphthalenesulfonate, sodium naphthalene-6-sulfonate, 2-naphthalene sulfonic acid sodium salt, sodium beta-naphthalenesulfonate, sodium naphthalene-2-sulphonate | |
| [Na+].[O-]S(=O)(=O)C1=CC=C2C=CC=CC2=C1 | |
| 230.21 | |
| 230.22 |
| 532-02-5 | |
| C10H7NaO3S | |
| 1000 g | |
| Hygroscopic | |
| YWPOLRBWRRKLMW-UHFFFAOYSA-M | |
| sodium naphthalene-2-sulfonate | |
| 23661868 | |
| 98% |
Chemical Identifiers
| 532-02-5 | |
| 230.21 | |
| YWPOLRBWRRKLMW-UHFFFAOYSA-M | |
| 23661868 |
| C10H7NaO3S | |
| MFCD00064186 | |
| sodium 2-naphthalenesulfonate, sodium naphthalene-2-sulfonate, 2-naphthalenesulfonic acid sodium salt, 2-naphthalenesulfonic acid, sodium salt, unii-r5f0ctd2oj, sodium-2-naphthalenesulfonate, sodium naphthalene-6-sulfonate, 2-naphthalene sulfonic acid sodium salt, sodium beta-naphthalenesulfonate, sodium naphthalene-2-sulphonate | |
| [Na+].[O-]S(=O)(=O)C1=CC=C2C=CC=CC2=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-523-8
RTECSNumber : QK3678000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only