missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega NanoBRET TE BET BRD DNA Bundle, 1 each
Description
- For Use With All NanoBRET™ TE Assays
- DNA Bundles and Transfection Carrier DNA
- Intracellular TE Nano−Glo(R) Substrate/Inhibitor
- Tracer Dilution Buffer
Specifications
Specifications
| Content And Storage | -30°C to -10°C |
| Quantity | 1 Each |