missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-tert-Butoxycarbonyl-Ngamma-trityl-L-asparagine 98.0+%, TCI America™

Catalog No. p-7123168
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37895G 5 g
B378925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37895G Supplier TCI America Supplier No. B37895G
Only null left

Chemical Identifiers

CAS 132388-68-2
Molecular Formula C28H30N2O5
Molecular Weight (g/mol) 474.557
MDL Number MFCD00153299
InChI Key PYGOCFDOBSXROC-QHCPKHFHSA-N
Synonym boc-asn trt-oh, nalpha-boc-ngamma-trityl-l-asparagine, nalpha-tert-butoxycarbonyl-ngamma-trityl-l-asparagine, s-2-tert-butoxycarbonyl amino-4-oxo-4-tritylamino butanoic acid, boc-n-beta-trityl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-triphenylmethylcarbamoyl propanoic acid, boc-ngamma-trityl-d-asparagine, boc-asn-trt-oh, pubchem18974, n, a-boc-n, a-trityl-l-asparagine
PubChem CID 11385960
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(tritylamino)butanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)NC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O

Specifications

Melting Point 202°C
Color White
Quantity 5 g
Formula Weight 474.56
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-tert-Butoxycarbonyl-Ngamma-trityl-L-asparagine