accessibility menu, dialog, popup

UserName

CAS RN 132388-68-2

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-[(triphenylmethyl)carbamoyl]propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(triphenylmethylcarbamoyl)propanoic acid
Molecular Weight (g/mol): 474.56
Molecular Formula: C28H30N2O5
InChi Key: PYGOCFDOBSXROC-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)