missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-tert-Butoxycarbonyl-Ngamma-trityl-L-asparagine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 202°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 474.56 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nalpha-tert-Butoxycarbonyl-Ngamma-trityl-L-asparagine |
Safety and Handling
| TSCA | No |