missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Nepsilon-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]-L-lysine, 95%, Thermo Scientific Chemicals

$134.00 - $398.96

Chemical Identifiers

CAS 204777-78-6
Molecular Formula C34H42N2O6
Molecular Weight (g/mol) 574.72
MDL Number MFCD01631658
InChI Key PYCBVLUBTMHNPW-MHZLTWQESA-N
Synonym fmoc-lys ivdde-oh, n, fmoc-l-lys ivdde-oh, a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene-3-methylbutyl-l-lysine, n-fmoc-n'-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene-3-methylbutyl-l-lysine, fmoc-lys ddiv-oh, fmoc-l-lys ivdde-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-1-4,4-dimethyl-2,6-dioxocyclohexylidene-3-methylbutyl amino hexanoic acid, a-fmoc-n-, a-1-4,4-dimethyl-2,6-dioxocyclohex-1, fmoc-lys ddiv-oh hplc, n-, a-fmoc-n-, a-1-4,4-dimethyl-2,6-dioxocyclohex-1
PubChem CID 11071922
IUPAC Name (2S)-6-{[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
SMILES CC(C)CC(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66820MD
View Documents
Thermo Scientific Chemicals
H66820MD
250 mg
Each for $134.00
Only null left
 
AAH6682003
View Documents
Thermo Scientific Chemicals
H6682003
1 g
Each for $398.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

204777-78-6
574.72
PYCBVLUBTMHNPW-MHZLTWQESA-N
11071922
CC(C)CC(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O
C34H42N2O6
MFCD01631658
fmoc-lys ivdde-oh, n, fmoc-l-lys ivdde-oh, a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene-3-methylbutyl-l-lysine, n-fmoc-n'-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene-3-methylbutyl-l-lysine, fmoc-lys ddiv-oh, fmoc-l-lys ivdde-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-1-4,4-dimethyl-2,6-dioxocyclohexylidene-3-methylbutyl amino hexanoic acid, a-fmoc-n-, a-1-4,4-dimethyl-2,6-dioxocyclohex-1, fmoc-lys ddiv-oh hplc, n-, a-fmoc-n-, a-1-4,4-dimethyl-2,6-dioxocyclohex-1
(2S)-6-{[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid

Specifications

204777-78-6
C34H42N2O6
fmoc-lys ivdde-oh, n, fmoc-l-lys ivdde-oh, a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene-3-methylbutyl-l-lysine, n-fmoc-n'-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene-3-methylbutyl-l-lysine, fmoc-lys ddiv-oh, fmoc-l-lys ivdde-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-1-4,4-dimethyl-2,6-dioxocyclohexylidene-3-methylbutyl amino hexanoic acid, a-fmoc-n-, a-1-4,4-dimethyl-2,6-dioxocyclohex-1, fmoc-lys ddiv-oh hplc, n-, a-fmoc-n-, a-1-4,4-dimethyl-2,6-dioxocyclohex-1
CC(C)CC(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O
250 mg
11071922
95%
Nalpha-Fmoc-Nepsilon-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]-L-lysine
White
MFCD01631658
PYCBVLUBTMHNPW-MHZLTWQESA-N
(2S)-6-{[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
574.72
574.72
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only