accessibility menu, dialog, popup

UserName

H3CCH3OOH3CCH3NH(S)OHONHOO

CAS RN 204777-78-6

IUPAC Name: (2S)-6-{[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2S)-6-{[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid|FMOC-LYS(IVDDE)-OH
Molecular Weight (g/mol): 574.72
Molecular Formula: C34H42N2O6
InChi Key: PYCBVLUBTMHNPW-MHZLTWQESA-N
SMILES: CC(C)CC(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)