missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Fmoc-Nepsilon-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]-L-lysine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6682003
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Nalpha-Fmoc-Nepsilon-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]-L-lysine | |
| White | |
| MFCD01631658 | |
| PYCBVLUBTMHNPW-MHZLTWQESA-N | |
| (2S)-6-{[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid | |
| 574.72 | |
| 574.72 | |
| Powder |
| 204777-78-6 | |
| C34H42N2O6 | |
| fmoc-lys ivdde-oh, fmoc-lys ddiv-oh, a-fmoc-n, a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene-3-methylbutyl-l-lysine, n-fmoc-n'-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene-3-methylbutyl-l-lysine, fmoc-lys ddiv-oh, fmoc-lys ddiv-oh hplc, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-1-4,4-dimethyl-2,6-dioxocyclohexylidene-3-methylbutyl amino hexanoic acid, a-fmoc-n-, a-1-4,4-dimethyl-2,6-dioxocyclohex-1, fmoc-lys ddiv-oh hplc, n-, a-fmoc-n-, a-1-4,4-dimethyl-2,6-dioxocyclohex-1 | |
| CC(C)CC(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O | |
| 1 g | |
| 11071922 | |
| 95% |
Chemical Identifiers
| 204777-78-6 | |
| 574.72 | |
| PYCBVLUBTMHNPW-MHZLTWQESA-N | |
| 11071922 | |
| CC(C)CC(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O |
| C34H42N2O6 | |
| MFCD01631658 | |
| fmoc-lys ivdde-oh, fmoc-lys ddiv-oh, a-fmoc-n, a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene-3-methylbutyl-l-lysine, n-fmoc-n'-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene-3-methylbutyl-l-lysine, fmoc-lys ddiv-oh, fmoc-lys ddiv-oh hplc, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-1-4,4-dimethyl-2,6-dioxocyclohexylidene-3-methylbutyl amino hexanoic acid, a-fmoc-n-, a-1-4,4-dimethyl-2,6-dioxocyclohex-1, fmoc-lys ddiv-oh hplc, n-, a-fmoc-n-, a-1-4,4-dimethyl-2,6-dioxocyclohex-1 | |
| (2S)-6-{[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only