missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Ngamma-trityl-L-asparagine, 98%

Catalog No. AAH6244603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6244603 1 g
AAH6244606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6244603 Supplier Thermo Scientific Chemicals Supplier No. H6244603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 132388-68-2 | C28H30N2O5 | 474.557 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132388-68-2
Molecular Formula C28H30N2O5
Molecular Weight (g/mol) 474.557
MDL Number MFCD00153299
InChI Key PYGOCFDOBSXROC-QHCPKHFHSA-N
Synonym boc-asn trt-oh, nalpha-boc-ngamma-trityl-l-asparagine, nalpha-tert-butoxycarbonyl-ngamma-trityl-l-asparagine, s-2-tert-butoxycarbonyl amino-4-oxo-4-tritylamino butanoic acid, boc-n-beta-trityl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-triphenylmethylcarbamoyl propanoic acid, boc-ngamma-trityl-d-asparagine, boc-asn-trt-oh, pubchem18974, n, a-boc-n, a-trityl-l-asparagine
PubChem CID 11385960
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(tritylamino)butanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)NC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O

Specifications

Melting Point 202°C to 205°C
Quantity 1 g
Beilstein 4339761
Formula Weight 474.55
Percent Purity 98%
Chemical Name or Material Nalpha-Boc-Ngamma-trityl-L-asparagine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only