missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Ngamma-trityl-L-asparagine 98.0+%, TCI America™

Catalog No. p-7129719
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05081G 1 g
F05085G 5 g
F050825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F05081G Supplier TCI America Supplier No. F05081G
Only null left

Chemical Identifiers

CAS 132388-59-1
Molecular Formula C38H31N2O5
Molecular Weight (g/mol) 595.68
MDL Number MFCD00077049
InChI Key KJYAFJQCGPUXJY-UMSFTDKQSA-M
Synonym fmoc-asn trt-oh, nalpha-fmoc-ngamma-trityl-l-asparagine, ambotzfaa1015, n-fmoc-n4-trityl-l-asparagine, pubchem9947, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-oxo-4-tritylamino butanoic acid, fmoc-n'-trityl-l-asparagine, nalpha-9h-fluoren-9-ylmethoxy carbonyl-ngamma-trityl-l-asparagine, n-9-fluorenyl methoxy carbonyl-n-trityl-l-asparagine
PubChem CID 640248
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[(triphenylmethyl)carbamoyl]propanoate
SMILES [O-]C(=O)[C@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White
Quantity 1 g
Formula Weight 596.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Ngamma-trityl-L-asparagine