missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-tetradecanoyl-L-lysine 98.0+%, TCI America™

Catalog No. p-7130081
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11431G 1 g
F11435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11431G Supplier TCI America Supplier No. F11431G

Chemical Identifiers

CAS 1128181-23-6
Molecular Formula C35H50N2O5
Molecular Weight (g/mol) 578.794
InChI Key PNXSYZSEFDFTDZ-YTTGMZPUSA-N
Synonym Nalpha-Fmoc-Nepsilon-tetradecanoyl-L-lysine, Fmoc-Lys(myristoyl)-OH, Fmoc-Lys(Myr)-OH
PubChem CID 25231984
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(tetradecanoylamino)hexanoic acid
SMILES CCCCCCCCCCCCCC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 140°C
Color White
Quantity 1 g
Formula Weight 578.79
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-tetradecanoyl-L-lysine